5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-(benzofuran-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, Benzofuran, 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-benzofuran
B5823-1G
B5823-250MG
TCB5823-1GEA
195
USD
TCB5823-1G
TCB5823-250MG
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan ≥98.0% (by GC, titration analysis)
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan
Formula:
C₁₄H₁₇BO₃ MW: 244.1 g/mol |
MDL Number:
MFCD04115374 CAS Number: 519054-55-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 70.0 to 74.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...