4'-Chlorobiphenyl-4-carboxaldehyde ≥98.0% (by GC)

Supplier: TCI America

Synonyms: [1,1'-Biphenyl]-4-carboxaldehyde, 4'-chloro-, 4'-Chlorobiphenyl-4-carbaldehyde, 4'-Chloro-4-biphenylcarbaldehyde, 4'-Chloro-[1,1'-biphenyl]-4-carboxaldehyde

C3616-5G C3616-1G
TCC3616-5GEA 162 USD
TCC3616-5G TCC3616-1G
4'-Chlorobiphenyl-4-carboxaldehyde ≥98.0% (by GC)
4'-Chlorobiphenyl-4-carboxaldehyde
Formula: C₁₃H₉ClO
MW: 216.67 g/mol
MDL Number: MFCD01631911
CAS Number: 80565-30-6

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 114.0 to 118.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR