1,3,5-Triformylbenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1,3,5-benzenetricarbaldehyde, Trimesic formaldehyde, Benzene-1,3,5-tricarbaldehyde

B6003-1G B6003-200MG
TCB6003-1GEA 150 USD
TCB6003-1G TCB6003-200MG
1,3,5-Triformylbenzene ≥98.0% (by GC)
1,3,5-Triformylbenzene
Formula: C₉H₆O₃
MDL Number: MFCD00182470
CAS Number: 3163-76-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 165.0 to 169.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR