N-(tert-Butoxycarbonyl)-L-alanine N'-methoxy-N'-methylamide ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (S)-TERT-BUTYL 1-(METHOXY(METHYL)AMINO)-1-OXOPROPAN-2-YLCARBAMATE, BOC-L-ALANINE N-METHOXY-N-METHYL AMIDE, (2S)-2-[(tert-butoxy)carbonylamino]-N-methoxy-N-methylpropanamide, Boc-Ala-NMe(OMe)

B6019-1G B6019-5G
TCB6019-1GEA 35 USD
TCB6019-1G TCB6019-5G
N-(tert-Butoxycarbonyl)-L-alanine N'-methoxy-N'-methylamide ≥98.0% (by GC)
N-(tert-Butoxycarbonyl)-L-alanine N'-methoxy-N'-methylamide
Formula: C₁₀H₂₀N₂O₄
MW: 232.28 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00151859
CAS Number: 87694-49-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 149.0 to 153.0 deg-C
Specific rotation [a]20/D -27.0 to -31.0 deg (C=1, methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR