Thiophene-2,5-diboronic acid bis(pinacol) ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B5871-1G B5871-5G
TCB5871-1GEA 73 USD
TCB5871-1G TCB5871-5G
Thiophene-2,5-diboronic acid bis(pinacol) ester ≥98.0% (by GC, titration analysis)
Thiophene-2,5-diboronic acid bis(pinacol) ester
Formula: C₁₆H₂₆B₂O₄S
MDL Number: MFCD09800563
CAS Number: 175361-81-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 227.0 to 231.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR