5-Bromo-2-fluorobenzeneboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 5-Bromo-2-fluorophenylboronic acid, 5-Bromo-2-fluorobenzeneboronic acid

B5065-25G B5065-5G
TCB5065-25GEA 331 USD
TCB5065-25G TCB5065-5G
5-Bromo-2-fluorobenzeneboronic acid (contains varying amounts of Anhydride)
5-Bromo-2-fluorobenzeneboronic acid
Formula: C₆H₅BBrFO₂
MW: 218.82 g/mol
Melting Pt: 174 °C
MDL Number: MFCD05664307
CAS Number: 112204-57-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR