3-Diethylamino-1-propyne ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

D5393-25ML D5393-5ML
TCD5393-25MLEA 86 USD
TCD5393-25ML TCD5393-5ML
3-Diethylamino-1-propyne ≥98.0% (by GC, titration analysis)
3-Diethylamino-1-propyne
Formula: C₇H₁₃N
Boiling Pt: 120 °C
Flash Pt: 20 °C
MDL Number: MFCD00039881
CAS Number: 4079-68-9
EINECS: 223-804-5
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR