2-(2-Chlorophenyl)benzimidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(2-Chlorophenyl)-1H-benzo[d]imidazole

C3576-25G C3576-5G
TCC3576-25GEA 203 USD
TCC3576-25G TCC3576-5G
2-(2-Chlorophenyl)benzimidazole ≥98.0% (by GC, titration analysis)
2-(2-Chlorophenyl)benzimidazole
Formula: C₁₃H₉ClN₂
MW: 228.68 g/mol
MDL Number: MFCD00051632
CAS Number: 3574-96-7

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 232.0 to 234.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR