(R)-(-)-2-Chloro-1-phenylethanol ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: (R)-2-Chloro-1-phenylethanol

C3561-25G C3561-5G
TCC3561-25GEA 290 USD
TCC3561-25G TCC3561-5G
(R)-(-)-2-Chloro-1-phenylethanol ≥98.0% (by GC)
(R)-(-)-2-Chloro-1-phenylethanol
Formula: C₈H₉ClO
MW: 156.6095 g/mol
MDL Number: MFCD00077691
CAS Number: 56751-12-3

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR