Ciclopirox olamine ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 6-Cyclohexyl-1-hydroxy-4-methylpyridin-2(1H)-one 2-aminoethanol salt

C3545-1G C3545-5G
TCC3545-1GEA 42 USD
TCC3545-1G TCC3545-5G
Ciclopirox olamine ≥98.0% (by HPLC)
Ciclopirox olamine
Formula: C₁₂H₁₇NO₂·C₂H₇NO
MW: 268.3520 g/mol
Melting Pt: 143 °C
MDL Number: MFCD00078997
CAS Number: 41621-49-2
EINECS: 255-464-9
Merck Index: 14,02266

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR