4,5-Dichloro-2-methyl-2H-pyridazin-3-one ≥98.0% (by GC)

Supplier: TCI America
Warning

D5324-1G D5324-5G
TCD5324-1GEA 24 USD
TCD5324-1G TCD5324-5G
4,5-Dichloro-2-methyl-2H-pyridazin-3-one ≥98.0% (by GC)
4,5-Dichloro-2-methyl-2H-pyridazin-3-one
Formula: C₅H₄Cl₂N₂O
MDL Number: MFCD00051686
CAS Number: 933-76-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 87.0 to 91.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR