1-Boc-5,6-dihydro-2H-pyridine-3-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 1-Boc-5,6-Dihydro-2H-pyridine-3-boronic acid Pinacol ester, tert-Butyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate, 1-(tert-Butoxycarbonyl)-1,2,5,6-tetrahydro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, tert-Butyl 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydropyridine-1(2H)-carboxylate, 2-(1-Boc-1,2,5,6-tetrahydro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Boc-1,2,5,6-tetrahydro-3-pyridylboronic acid Pinacol ester, 2-[1-(tert-Butoxycarbonyl)-1,2,5,6-tetrahydro-3-pyridyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Boc-1,2,5,6-tetrahydro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 1-(tert-Butoxycarbonyl)-1,2,5,6-tetrahydro-3-pyridylboronic acid Pinacol ester, 1-(tert-Butoxycarbonyl)-5,6-Dihydro-2H-pyridine-3-boronic acid Pinacol ester

B5844-1G B5844-200MG
TCB5844-1GEA 191 USD
TCB5844-1G TCB5844-200MG
1-Boc-5,6-dihydro-2H-pyridine-3-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
1-Boc-5,6-dihydro-2H-pyridine-3-boronic acid pinacol ester
Formula: C₁₆H₂₈BNO₄
MW: 309.22 g/mol
MDL Number: MFCD10697911
CAS Number: 885693-20-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 103.0 to 107.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR