tert-Butyldimethyl(2-propynyloxy)silane ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Tert-Butyldimethyl(2-propynyloxy)silane, tert-Butyldimethyl(prop-2-ynyloxy)silane

B5163-25ML B5163-5ML
TCB5163-25MLEA 176 USD
TCB5163-25ML TCB5163-5ML
tert-Butyldimethyl(2-propynyloxy)silane ≥98.0% (by GC)
tert-Butyldimethyl(2-propynyloxy)silane
Formula: C₉H₁₈OSi
Boiling Pt: 163 °C
MDL Number: MFCD28156589
CAS Number: 76782-82-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR