4-(Benzyloxy)benzyl chloride ≥98.0% (by GC)

Supplier: TCI America
Danger

B5121-1G B5121-5G
TCB5121-1GEA 100 USD
TCB5121-1G TCB5121-5G
4-(Benzyloxy)benzyl chloride ≥98.0% (by GC)
4-(Benzyloxy)benzyl chloride
Formula: C₁₄H₁₃ClO
MW: 232.71 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00000916
CAS Number: 836-42-0
EINECS: 212-648-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 77.0 to 81.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR