1-Benzyl-3,3-difluoro-4,4-piperidinediol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-Benzyl-3,3-difluoropiperidine-4,4-diol

B5930-1G B5930-200MG
TCB5930-1GEA 133 USD
TCB5930-1G TCB5930-200MG
1-Benzyl-3,3-difluoro-4,4-piperidinediol ≥98.0% (by GC, titration analysis)
1-Benzyl-3,3-difluoro-4,4-piperidinediol
Formula: C₁₂H₁₅F₂NO₂
MW: 243.26 g/mol
MDL Number: MFCD20926176
CAS Number: 1067914-81-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 121.0 to 125.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR