1,5-Dimethyl-3-(trifluoromethyl)-1H-pyrazole ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1,5-Dimethyl-3-(trifluoromethyl)pyrazole

D5071-1G D5071-200MG
TCD5071-1GEA 87 USD
TCD5071-1G TCD5071-200MG
1,5-Dimethyl-3-(trifluoromethyl)-1H-pyrazole ≥98.0% (by GC)
1,5-Dimethyl-3-(trifluoromethyl)-1H-pyrazole
Formula: C₆H₇F₃N₂
MW: 164.13 g/mol
Boiling Pt: 71 °C
MDL Number: MFCD04972844
CAS Number: 79080-31-2
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR