4-(Pyrimidin-2-yl)bromobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-(4-Bromophenyl)pyrimidine

B5806-1G B5806-200MG
TCB5806-1GEA 291 USD
TCB5806-1G TCB5806-200MG
4-(Pyrimidin-2-yl)bromobenzene ≥98.0% (by GC)
4-(Pyrimidin-2-yl)bromobenzene
Formula: C₁₀H₇BrN₂
MW: 235.09 g/mol
MDL Number: MFCD08056329
CAS Number: 210354-17-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 136.0 to 140.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR