5-Bromo-2-methylbenzyl alcohol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (5-Bromo-2-methylphenyl)methanol

B5037-1G B5037-5G
TCB5037-1GEA 180 USD
TCB5037-1G TCB5037-5G
5-Bromo-2-methylbenzyl alcohol ≥98.0% (by GC)
5-Bromo-2-methylbenzyl alcohol
Formula: C₈H₉BrO
MW: 201.07 g/mol
MDL Number: MFCD12031921
CAS Number: 258886-04-3

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 62.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR