(Dimethanesulfinylidene)amine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Dimethylsulphoximine, (S-Methylsulphonimidoyl)methane

D4971-1G D4971-5G
TCD4971-1GEA 85 USD
TCD4971-1G TCD4971-5G
(Dimethanesulfinylidene)amine ≥98.0% (by GC)
(Dimethanesulfinylidene)amine

Dimethylsulfoximine [D4971] is a building block for the synthesis of sulfoximines and utilized in various types of sulfoximidation reactions.


Possible applications include such as SN2 reactions, nucleophilic attacks to carbonyl groups and cross-coupling reactions, and providing the target compounds in high yields under various conditions. Furthermore, this functional group can be inserted at a late stage of complex syntheses via C-H activation.

Formula: C₂H₇NOS
MW: 93.15 g/mol
Boiling Pt: 100 °C
MDL Number: MFCD02656633
CAS Number: 1520-31-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR