2-Bromo-5-methylbenzaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

B5824-1G B5824-5G
TCB5824-1GEA 80 USD
TCB5824-1G TCB5824-5G
2-Bromo-5-methylbenzaldehyde ≥98.0% (by GC)
2-Bromo-5-methylbenzaldehyde
Formula: C₈H₇BrO
Boiling Pt: 100 °C
MDL Number: MFCD08669550
CAS Number: 90221-55-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 41.0 to 45.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR