2-Amino-5-chlorobenzothiazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-chloro-1,3-benzothiazol-2-amine

A3177-1G A3177-5G
TCA3177-1GEA 54 USD
TCA3177-1G TCA3177-5G
2-Amino-5-chlorobenzothiazole ≥98.0% (by HPLC, titration analysis)
2-Amino-5-chlorobenzothiazole
Formula: C₇H₅ClN₂S
MW: 184.65 g/mol
MDL Number: MFCD00205355
CAS Number: 20358-00-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 199.0 to 203.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR