4-Carboxyphenylacetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: (4-Carboxyphenyl)acetic acid, 4-(Carboxymethyl)benzoic acid

C3530-25G C3530-5G
TCC3530-25GEA 227 USD
TCC3530-25G TCC3530-5G
4-Carboxyphenylacetic acid ≥98.0% (by titrimetric analysis)
4-Carboxyphenylacetic acid
Formula: C₉H₈O₄
MDL Number: MFCD01074834
CAS Number: 501-89-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 243.0 to 247.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR