2,2-Difluoroethyl acetate ≥98.0% (by GC)

Supplier: TCI America
Danger

D5040-25G D5040-5G
TCD5040-25GEA 681 USD
TCD5040-25G TCD5040-5G
2,2-Difluoroethyl acetate ≥98.0% (by GC)
2,2-Difluoroethyl acetate
Formula: C₄H₆F₂O₂
Boiling Pt: 106 °C
MDL Number: MFCD00069133
CAS Number: 1550-44-3
UN: 3272
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR