(3-(Bromomethyl)-1-(p-toluenesulfonyl)azetidin-3-yl)methanol ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: [3-(Bromomethyl)-1-(p-toluenesulphonyl)azetidin-3-yl]methanol, [3-(Bromomethyl)-1-tosylazetidin-3-yl]methanol

B5852-1G B5852-200MG
TCB5852-1GEA 218 USD
TCB5852-1G TCB5852-200MG
(3-(Bromomethyl)-1-(p-toluenesulfonyl)azetidin-3-yl)methanol ≥98.0% (by GC)
(3-(Bromomethyl)-1-(p-toluenesulfonyl)azetidin-3-yl)methanol
Formula: C₁₂H₁₆BrNO₃S
MDL Number: MFCD18782897
CAS Number: 1041026-55-4

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 98.0 to 102.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR