tert-Butyl-7-oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate ≥95.0% (by GC)

Supplier: TCI America
Warning

B5969-1G B5969-5G
TCB5969-1GEA 67 USD
TCB5969-1G TCB5969-5G
tert-Butyl-7-oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate ≥95.0% (by GC)
tert-Butyl-7-oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate
Formula: C₁₀H₁₇NO₃
MDL Number: MFCD12407050
CAS Number: 161157-50-2

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR