2-(1-tert-Butoxycarbonylazetidin-3-yl)acetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-[1-Boc-azetidin-3-yl]acetic acid, 2-[1-(tert-Butoxycarbonyl)azetidin-3-yl]acetic acid

B6013-1G B6013-5G
TCB6013-1GEA 44 USD
TCB6013-1G TCB6013-5G
2-(1-tert-Butoxycarbonylazetidin-3-yl)acetic acid ≥98.0% (by GC, titration analysis)
2-(1-tert-Butoxycarbonylazetidin-3-yl)acetic acid
Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
MDL Number: MFCD02179028
CAS Number: 183062-96-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 116.0 to 120.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR