tert-Butyl-3-(2-ethoxy-2-oxoethylidene)azetidine-1-carboxylate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: tert-Butyl 3-(2-ethoxy-2-oxoethylidene)azetidine-1-carboxylate

B6012-1G B6012-200MG
TCB6012-1GEA 118 USD
TCB6012-1G TCB6012-200MG
tert-Butyl-3-(2-ethoxy-2-oxoethylidene)azetidine-1-carboxylate ≥98.0% (by GC)
tert-Butyl-3-(2-ethoxy-2-oxoethylidene)azetidine-1-carboxylate
Formula: C₁₂H₁₉NO₄
MW: 241.29 g/mol
MDL Number: MFCD16140210
CAS Number: 1002355-96-5

Order Now

Specification Test Results

Appearance White to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR