1,3-Dihydroxy-9H-xanthen-9-one ≥98.0% (by HPLC)

Supplier: TCI America

D5344-1G D5344-200MG
TCD5344-1GEA 191 USD
TCD5344-1G TCD5344-200MG
1,3-Dihydroxy-9H-xanthen-9-one ≥98.0% (by HPLC)
1,3-Dihydroxy-9H-xanthen-9-one
Formula: C₁₃H₈O₄
Melting Pt: 259 °C
MDL Number: MFCD00800670
CAS Number: 3875-68-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Carbon) 66.00 to 71.00 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR