6-Chloro-7-hydroxy-4-methyl-2H-chromen-2-one ≥98.0% (by GC)

Supplier: TCI America
Warning

C3319-1G C3319-5G
TCC3319-1GEA 118 USD
TCC3319-1G TCC3319-5G
6-Chloro-7-hydroxy-4-methyl-2H-chromen-2-one ≥98.0% (by GC)
6-Chloro-7-hydroxy-4-methyl-2H-chromen-2-one
Formula: C₁₀H₇ClO₃
Melting Pt: 280 °C
MDL Number: MFCD00816527
CAS Number: 19492-02-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR