6-Phenyl-4,5-dihydropyridazin-3(2H)-one ≥98.0% (by HPLC)

Supplier: TCI America
Warning

D5119-1G D5119-5G
TCD5119-1GEA 88 USD
TCD5119-1G TCD5119-5G
6-Phenyl-4,5-dihydropyridazin-3(2H)-one ≥98.0% (by HPLC)
6-Phenyl-4,5-dihydropyridazin-3(2H)-one
Formula: C₁₀H₁₀N₂O
MDL Number: MFCD00099597
CAS Number: 1011-46-7

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 150.0 to 154.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR