tert-Butyl-4-(2-ethoxy-2-oxoethylidene)tetrahydro-1(2H)-pyridinecarboxylate ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4-(2-Ethoxy-2-oxoethylidene)piperidine-1-carboxylic acid tert-Butyl ester, Ethyl [1-(tert-Butoxycarbonyl)piperidin-4-ylidene]acetate, tert-Butyl 4-(2-Ethoxy-2-oxoethylidene)piperidine-1-carboxylate, (1-Boc-piperidin-4-ylidene)acetic acid ethyl ester, [1-(tert-Butoxycarbonyl)piperidin-4-ylidene]acetic acid ethyl ester, Ethyl (1-Boc-piperidin-4-ylidene)acetate

B5855-1G B5855-5G
TCB5855-1GEA 45 USD
TCB5855-1G TCB5855-5G
tert-Butyl-4-(2-ethoxy-2-oxoethylidene)tetrahydro-1(2H)-pyridinecarboxylate ≥97.0% (by GC)
tert-Butyl-4-(2-ethoxy-2-oxoethylidene)tetrahydro-1(2H)-pyridinecarboxylate
Formula: C₁₄H₂₃NO₄
MW: 269.34 g/mol
MDL Number: MFCD12912655
CAS Number: 135716-08-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 83.0 to 87.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR