Boc-L-β-homophenylalanine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B6074-250MG B6074-1G
TCB6074-250MGEA 82 USD
TCB6074-250MG TCB6074-1G
Boc-L-β-homophenylalanine ≥98.0% (by HPLC, titration analysis)
Boc-L-beta-homophenylalanine
Formula: C₁₅H₂₁NO₄
MDL Number: MFCD01076271
CAS Number: 51871-62-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 104.0 to 108.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR