tert-Butyl-3-[(methylsulfonyl)oxy]-1-azetanecarboxylate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Boc-3-Methanesulphonyloxyazetidine

B5801-25G B5801-5G
TCB5801-25GEA 157 USD
TCB5801-25G TCB5801-5G
tert-Butyl-3-[(methylsulfonyl)oxy]-1-azetanecarboxylate ≥98.0% (by GC)
tert-Butyl-3-[(methylsulfonyl)oxy]-1-azetanecarboxylate
Formula: C₉H₁₇NO₅S
MW: 251.3 g/mol
MDL Number: MFCD05664832
CAS Number: 141699-58-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 67.0 to 71.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR