(2E)-3-(3-Aminophenyl)acrylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (E)-3-Aminocinnamic acid, (E)-3-(3-Aminophenyl)acrylic acid

A3297-1G A3297-5G
TCA3297-1GEA 103 USD
TCA3297-1G TCA3297-5G
(2E)-3-(3-Aminophenyl)acrylic acid ≥98.0% (by HPLC, titration analysis)
(2E)-3-(3-Aminophenyl)acrylic acid
Formula: C₉H₉NO₂
MW: 163.18 g/mol
MDL Number: MFCD00460112
CAS Number: 127791-53-1

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 180.0 to 184.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR