7-Methoxy-3,4,5,6-tetrahydro-2H-azepine ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
1-Aza-2-methoxy-1-cycloheptene, O-Methylcaprolactim, 7-Methoxy-3,4,5,6-tetrahydro-2H-azepine
A3282-25G
A3282-5G
TCA3282-25GEA
99
USD
TCA3282-25G
TCA3282-5G
7-Methoxy-3,4,5,6-tetrahydro-2H-azepine ≥98.0% (by GC)
7-Methoxy-3,4,5,6-tetrahydro-2H-azepine
Formula:
C₇H₁₃NO MW: 127.19 g/mol Boiling Pt: 53 °C Flash Pt: 51 °C |
MDL Number:
MFCD00006938 CAS Number: 2525-16-8 EINECS: 219-762-2 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...