3-Bromo-5-nitropyridin-4-ol ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
B5287-1G
B5287-5G
TCB5287-1GEA
133
USD
TCB5287-1G
TCB5287-5G
3-Bromo-5-nitropyridin-4-ol ≥98.0% (by titrimetric analysis)
3-Bromo-5-nitropyridin-4-ol
Formula:
C₅H₃BrN₂O₃ |
MDL Number:
MFCD08692339 CAS Number: 31872-65-8 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...