1-(4-Isocyanatophenyl)ethanone ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Isocyanic acid 4-Acetylphenyl ester, 4-Acetylphenylisocyanate, 4-Acetylphenyl Isocyanate

A2933-1G A2933-5G
TCA2933-1GEA 47 USD
TCA2933-1G TCA2933-5G
1-(4-Isocyanatophenyl)ethanone ≥98.0% (by GC)
1-(4-Isocyanatophenyl)ethanone
Formula: C₉H₇NO₂
Boiling Pt: 139 °C
Melting Pt: 37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013879
CAS Number: 49647-20-3

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR