2-Acetylamino-4-methylthiazole-5-sulfonyl chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
2-Acetylamino-4-methyl-thiazole-5-sulphonyl chloride, 2-Acetamido-4-methyl-1,3-thiazole-5-sulphonyl chloride, 2-Acetamido-4-methylthiazole-5-sulphonyl chloride, 2-Acetamido-4-methyl-5-thiazolesulphonyl chloride
A3288-1G
A3288-5G
TCA3288-1GEA
57
USD
TCA3288-1G
TCA3288-5G
2-Acetylamino-4-methylthiazole-5-sulfonyl chloride ≥98.0% (by HPLC, titration analysis)
2-Acetylamino-4-methylthiazole-5-sulfonyl chloride
Formula:
C₆H₇ClN₂O₃S₂ |
MDL Number:
MFCD00005320 CAS Number: 69812-29-9 EINECS: 274-127-7 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 155.0 to 159.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...