2,2-Difluoroethyl trifluoromethanesulfonate ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 2,2-Difluoroethyltrifluoromethanesulphonate

D5299-1G D5299-5G
TCD5299-1GEA 46 USD
TCD5299-1G TCD5299-5G
2,2-Difluoroethyl trifluoromethanesulfonate ≥98.0% (by GC)
2,2-Difluoroethyl trifluoromethanesulfonate
Formula: C₃H₃F₅O₃S
MW: 214.11 g/mol
Boiling Pt: 118 °C
MDL Number: MFCD02093342
CAS Number: 74427-22-8
UN: 2920
ADR: 8 II

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR