3-(1H-Indol-3-yl)-3-oxopropanenitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 3-(Cyanoacetyl)indole

C3197-1G C3197-5G
TCC3197-1GEA 172 USD
TCC3197-1G TCC3197-5G
3-(1H-Indol-3-yl)-3-oxopropanenitrile ≥98.0% (by GC)
3-(1H-Indol-3-yl)-3-oxopropanenitrile
Formula: C₁₁H₈N₂O
MW: 184.2 g/mol
MDL Number: MFCD04610443
CAS Number: 20356-45-0
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR