2-Bromo-1-(1-pyrrolidinyl)-1-ethanone ≥93.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-(Bromoacetyl)pyrrolidine, 2-Bromo-1-(pyrrolidin-1-yl)ethanone

B5307-1G B5307-200MG
TCB5307-1GEA 193.56 USD
TCB5307-1G TCB5307-200MG
2-Bromo-1-(1-pyrrolidinyl)-1-ethanone ≥93.0% (by GC)
2-Bromo-1-(1-pyrrolidinyl)-1-ethanone
Formula: C₆H₁₀BrNO
MW: 192.06 g/mol
Boiling Pt: 120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06800371
CAS Number: 90892-09-4

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 93.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR