2-Amino-4-phenylpyrimidine ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Phenylpyrimidin-2-amine

A3244-1G A3244-5G
TCA3244-1GEA 55 USD
TCA3244-1G TCA3244-5G
2-Amino-4-phenylpyrimidine ≥97.0% (by GC, titration analysis)
2-Amino-4-phenylpyrimidine
Formula: C₁₀H₉N₃
MW: 171.2 g/mol
MDL Number: MFCD00094706
CAS Number: 2305-87-5

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 162.0 to 166.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR