(3-Chloro-4-methoxyphenyl)methanamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

C3634-1G C3634-5G
TCC3634-1GEA 43 USD
TCC3634-1G TCC3634-5G
(3-Chloro-4-methoxyphenyl)methanamine ≥98.0% (by GC, titration analysis)
(3-Chloro-4-methoxyphenyl)methanamine
Formula: C₈H₁₀ClNO
Boiling Pt: 110 °C
MDL Number: MFCD00995395
CAS Number: 115514-77-7
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR