3-Bromo-4,5-dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B5252-1G B5252-5G
TCB5252-1GEA 159 USD
TCB5252-1G TCB5252-5G
3-Bromo-4,5-dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
3-Bromo-4,5-dimethoxybenzoic acid
Formula: C₉H₉BrO₄
MDL Number: MFCD00094247
CAS Number: 20731-48-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 193.0 to 197.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR