Boc-L-His(Bzl)-OH ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Boc-His(Bzl)-OH, tele-Benzyl-Nalpha-(tert-butoxycarbonyl)-L-histidine

B5735-1G B5735-5G
TCB5735-1GEA 73 USD
TCB5735-1G TCB5735-5G
Boc-L-His(Bzl)-OH ≥98.0% (by titrimetric analysis)
Boc-L-His(Bzl)-OH
Formula: C₁₈H₂₃N₃O₄
MW: 345.4 g/mol
Melting Pt: 182 °C
MDL Number: MFCD00066093
CAS Number: 20898-44-6
EINECS: 244-105-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Specific rotation [a]20/D +22.0 to +25.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR