Boc-Aib-ol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: N-Boc-2-amino-2-methyl-1-propanol, N-tert-Butoxycarbonyl-2-amino-2-methyl-1-propanol, tert-Butyl (1-Hydroxy-2-methylpropan-2-yl)carbamate

B6025-1G B6025-5G
TCB6025-1GEA 28 USD
TCB6025-1G TCB6025-5G
Boc-Aib-ol ≥98.0% (by GC)
Boc-Aib-ol
Formula: C₉H₁₉NO₃
MW: 189.3 g/mol
MDL Number: MFCD03788641
CAS Number: 102520-97-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 59.0 to 63.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR