Ac-β-Ala-OH ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: N-Ac-β-Ala-OH, 3-Acetamidopropanoic acid, N-Acetyl-beta-alanine, N-Acetyl-b-alanine

A3205-25G A3205-5G
TCA3205-25GEA 217 USD
TCA3205-25G TCA3205-5G
Ac-β-Ala-OH ≥98.0% (by titrimetric analysis)
Ac-β-Ala-OH
Formula: C₅H₉NO₃
MW: 131.1 g/mol
MDL Number: MFCD00037296
CAS Number: 3025-95-4
EINECS: 221-185-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 79.0 to 83.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR