Benztropine mesylate ≥98.0% (by HPLC)

Supplier: TCI America
Danger

B5592-25G B5592-5G
TCB5592-25GEA 422 USD
TCB5592-25G TCB5592-5G
Benztropine mesylate ≥98.0% (by HPLC)
Benztropine mesylate
Formula: C₂₂H₂₉NO₄S
Storage Temperature: Refrigerator
MDL Number: MFCD00074784
CAS Number: 132-17-2
EINECS: 205-048-8
UN: 2811
ADR: 6.1,III
Merck Index: 14,01121

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Melting point 139.0 to 143.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR