Boc-L-cyclopropylglycine

Supplier: TCI America

B6088-1G B6088-200MG
TCB6088-1GEA 163 USD
TCB6088-1G TCB6088-200MG
Boc-L-cyclopropylglycine
Boc-L-cyclopropylglycine
Formula: C₁₀H₁₇NO₄
Melting Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659132
CAS Number: 155976-13-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Melting point 116.0 to 120.0 deg-C
Specific rotation [a]20/D +35.0 to +41.0 deg (C=1, ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR