1-Bromo-3-chloro-2-iodobenzene ≥97.0% (by GC)

Supplier: TCI America
Warning

B5765-1G B5765-5G
TCB5765-1GEA 46 USD
TCB5765-1G TCB5765-5G
1-Bromo-3-chloro-2-iodobenzene ≥97.0% (by GC)
1-Bromo-3-chloro-2-iodobenzene
Formula: C₆H₃BrClI
MDL Number: MFCD09907935
CAS Number: 450412-28-9

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 77.0 to 81.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR